Cryogenic Automation Labels for Barcode Labelers
Supplier: GA INTERNATIONAL INC
Certificates
About this item
Cryogenic automation labels, compatible with most automated print-and-apply systems. These labels are designed to withstand long-term storage in liquid nitrogen (−196 °C) and laboratory freezers (−80 °C) and can be printed with all major thermal transfer automated labeling systems.
- Temperature range: −196 to +100 °C (−320.8 to +212 °F)
- Resists cryogenic temperature storage including immersion in liquid nitrogen
- Compatible with all automated print-and-apply printers and labelers
Cryogenic automation labels are designed for use with automated print-and-apply barcode systems for tubes and cryo vials. These waterproof labels withstand long-term storage in liquid nitrogen (−196 °C/−321 °F), and laboratory freezers (−80 °C/−112 °F). Customized for use with any major brand of thermal-transfer automated labeling system.
Frequently Bought Together