Fetal Bovine Serum, Premium, Australia Origin (Heat Inactivated and Gamma Irradiated)
Catalog # 76339-832
Supplier: Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Description:Fetal bovine serum with weight 1.4 lbs.
- Size:500 ml
- Environmentally Preferable:
- Origin:AU
- Storage Temperature:−4...−10 °C
- Cat. No.:76339-832
Specifications
About this item
Species: Bovine, bos taurus
Type: Fetal
Treatment: Heat inactivated and gamma irradiated
Format: Liquid
Shelf life: 60 months
- Collected at government registered facilities in Australia
- Exported from the country of origin accompanied with a veterinarian health certificate and imported under a USDA-APHIS issued import permit
- True pooled and is triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
- Validated process used to heat inactivate serum at 56 °C (±2 °C) for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath
- Validated process used to gamma irradiate serum within one of two separate dose ranges: 25 to 40 kGy and 30 to 45 kGy
Certifications: Certificate of Suitability, R0-CEP 2020-127, by the European Directorate for the Quality of Medicines (EDQM).