Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy3 phosphoramidite
Cy3 phosphoramidite
Cy3 phosphoramidite
Cy3 phosphoramidite
Catalog # 77462-964
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
Cy3 phosphoramidite
Catalog # 77462-964
Supplier:
AAT BIOQUEST INC
Supplier Number:
6070
CAS Number:
Specifications
Size:
100 µmoles
Environmentally Preferable:
Cat. No.:
77462-964
Specifications
About this item
Product Details
:
:
953.63 g/mol
CAS Number:
182873-76-3
Product Details