ContactUsLinkComponent
IVD-US Spot Saver Card
IVD-US Spot Saver Card
Catalog # 77569-990
New Product
IVD-US Spot Saver Card
Catalog # 77569-990
Supplier Number:  GRNK11022
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    IVD-US Spot saver card
  • Environmentally Preferable:
  • Cat. No.:
    77569-990
  • Packaging:
    100/case
Specifications
About this item
GRNK1 cards are a medium for the collection and transportation of blood specimen spots to a laboratory, incorporating the high quality Ahlstrom 226 grade paper which ensures even and uniform sample distribution.
  • Two part card
  • 226 filter paper, 125# white tag and flap to cover blood spots
  • Code 3 of 9 Barcode with one human readable (serial number)
  • Blood collection circles Int.Ø = 12.7 mm
  • Final folded card size: 3²/₃(W) x 2⁵/₈"(H)
For in vitro diagnostic use.