Order Entry
United States
ContactUsLinkComponent
Mouse IL-12 (p70) ELISA Kit
Mouse IL-12 (p70) ELISA Kit
Catalog # MSPP-02026
CAS Number:  
undefined
Mouse IL-12 (p70) ELISA Kit
Catalog # MSPP-02026
Supplier Number:  02026
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Interleukin-12
  • Size:
    2 units
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-02026

Specifications

About this item

For detection and measurement of mouse interleukin 12 (p70) subunit.

The Mouse Interleukin 12 (IL-12) p70 ELISA Kit is designed for the quantitative detection and measurement of mouse IL-12 (p70) in biological fluids such as serum, plasma, and cell culture supernatants. IL-12 (p70) is a heterodimeric cytokine composed of p35 and p40 subunits; the p40 subunit is shared with IL-23. IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells.

IL-12 is a regulator of cytokine synthesis, especially IFN-γ, enhances the proliferation of T and NK cells, and stimulates the differentiation of CD⁴⁺ and CD⁸⁺ T cells. The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine.

The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’, 5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample.

The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').