Order Entry
United States
ContactUsLinkComponent
Methylene blue solution in methanol for cell staining
Methylene blue solution in methanol for cell staining
Catalog # TCM2392-100ML
Supplier:  TCI America
New Product
CAS Number:  
Methylene blue solution in methanol for cell staining
Catalog # TCM2392-100ML
Supplier:  TCI America
Supplier Number:  M2392-100ML
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    TCM2392-100ML

Specifications

Specification Test Results

  • Appearance:
    Blue to Dark blue clear liquid to cloudy liquid
  • Absorbance:
    min. 350 (664 nm)
  • Cell staining test:
    Extent of staining same as reference

Specification Test Results

About this item

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
 :
 :319.86 g/mol
EINECS:200-515-2
UN:1230
 :GHS02
 :GHS07
 :GHS08
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008
Melting Pt:180
ADR:3 II
Merck Index:14,06060