Order Entry
United States
ContactUsLinkComponent
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Catalog # MSPP-60067
undefined
Anti-TRA-2-54 Mouse Monoclonal Antibody [clone: TRA-2-54/2J]
Catalog # MSPP-60067
Supplier Number:  60067
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen Name:
    TRA-2-54
  • Clonality:
    Monoclonal
  • Clone:
    TRA-2-54/2J
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Rabbit,
    Non-human primates,
    Chimpanzee,
    Human,
    Cat
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    249
  • Cat. No.:
    MSPP-60067

Specifications

About this item

Mouse monoclonal IgG1 antibody against human, chimpanzee, gibbon TRA-2-54 (tissue non-specific alkaline phosphatase), unconjugated.

  • Pluripotent Stem cells

The TRA-2-54/2J antibody reacts with the tissue non-specific isoform of alkaline phosphatase (AP), which is expressed at high levels in undifferentiated pluripotent stem cells such as human and mouse embryonic stem (ES) cells, as well as induced pluripotent stem (iPS) and embryonal carcinoma (EC) cells. This isozyme of AP is the same as that expressed by liver, bone and kidney cells. Levels of AP decrease upon differentiation and, thus, the TRA-2-54/2J antibody can be used to monitor the differentiation status of pluripotent stem cells. The TRA-2-54/2J antibody does not cross-react with other isozymes of AP, which differ through post-translational modification, and reportedly does not recognize AP from cow, dog, goat, Guinea pig, hamster, mouse, rat or sheep, or Old World Monkeys such as the rhesus monkey, baboon and African green monkey.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: TRA-2-54
Clonality: Monoclonal
Clone: TRA-2-54/2J
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cat;Chimpanzee;Human;Non-Human Primate;Pig;Rabbit