Order Entry
United States
ContactUsLinkComponent
Mouse Insulin ELISA
Mouse Insulin ELISA
Catalog # 101720-326
Supplier:  Alpco Diagnostics
CAS Number:  
Mouse Insulin ELISA
Catalog # 101720-326
Supplier:  Alpco Diagnostics
Supplier Number:  80-INSMS-E01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Insulin
  • Description:
    Mouse insulin ELISA
  • Size:
    1×96 wells
  • Environmentally Preferable:
  • Sample Type:
    Plasma, serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h, 15 min
  • Detection Range:
    0.188 - 6.9 ng/ml
  • Sample Volume:
    10 µl
  • Sensitivity:
    0.06 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101720-326
  • Supplier no.:
    80-INSMS-E01

Specifications

About this item

The mouse insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes with just 10 µl of serum or plasma sample.

  • Low sample volume of 10 µl
  • Dynamic range of 0.188 to 6.9 ng/ml
  • Superior sensitivity of 0.06 ng/ml

Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the mouse insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time.

Caution: For research use only. Not for use in diagnostic procedures.