Order Entry
United States
ContactUsLinkComponent
VWR® Traceable® Digital Conductivity Bench Meter
VWR® Traceable® Digital Conductivity Bench Meter
Catalog # 61161-362
Supplier:  VWR International
undefined
VWR® Traceable® Digital Conductivity Bench Meter
Catalog # 61161-362
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Temperature Range:
    –30 to 130°C (–34 to 266°F)
  • Description:
    Conductivity bench meter
  • Environmentally Preferable:
  • Conductivity Accuracy:
    ±0.3% plus 1 digit
  • Conductivity Range:
    0.01 to 20.00 µS, 0.1 to 200 µS, 1 to 2000 µS, 10 to 20,000 µS, 100 to 200,000 µS, 0.001 to 2.00 megohms, 0.01 to 20.00 megohms, 0.1 to 20.00 ppm dissolved solids, 1 to 200 ppm dissolved solids, 10 to 2000 ppm dissolved solids, 100 to 20,000 ppm dissolved solids
  • Dimensions:
    15.24L x 20.95H x 8.89D cm (6 x 81/4 x 31/2")
  • Probe:
    1.27 dia. x 15.24L cm (1/2 x 6")
  • Weight:
    0.68 kg (1.5 lbs.)
  • Cat. No.:
    61161-362

Specifications

About this item

Designed to measure conductivity in water analysis, biotechnology, life sciences, environmental studies, and more.

  • Traceable®
  • VWR® one-year limited warranty

Can also be used to check water purity in stills and demineralizers, test laboratory glassware, measure total dissolved solids, and make up solutions.

Large, 4-digit, 12.7 mm (¹/₂") LCD shows temperature in °C and °F. Instant-response probe contains platinum electrodes and a solid-state thermometer for automatic/manual temperature compensation. All readings are referenced to the international standard of 25 °C. Computer RS232 output and recorder jack allows for continuous monitoring and a permanent record. Unit has keys to calibrate to four different calibration points. Chemical-resistant ABS housing withstands harsh environments.

Accurately provides results in conductivity (micromhos/cm and microsiemens/cm), resistivity (megohms), and dissolved solids (parts per million).

Certifications: Meets all accreditation requirements and U.S. Government specifications. Measures reagent-grade water for CAP, ASTM, NCCLS, and ACS. A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with glass probe, 115 V AC adapter, 9 V battery, receptacle to supply probe replatinizing current, and probe holder arm.