Specifications
- Color:Clear
- Description:Storage Plate with Optimized Geometry for Low Evaporation
- Environmentally Preferable:
- Cat. No.:89085-692
- Working volume:1 - 14 µl
- Supplier no.:792870-906
- Total Volume:16 µl
Specifications
About this item
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination.
- Solid bottom design
- Nonsterile
- Free of DNase, RNase, and human DNA
- Designed for low evaporation
The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range from a starting plate into a target plate by means of ultrasound
All plates are manufactured from cycloolefin and feature solid bottoms and F-bottom well profiles. The LoBase storage plate is ideal for working with minimal sample volumes. The storage plate with optimized geometry for low evaporation is equipped with a groove around the plate edges for a matching cycloolefin plate lid (not included). This prevents evaporation and minimizes edge effects. All plates are nonsterile, nonpyrogenic, and DNase-, RNase-, and human DNA-free.
The plates used have a substantial influence on the accuracy of the amount of liquid transferred. Even wall thicknesses and consistent sealing of the microplate bottoms are essential in maintaining the reproducibility of the sample.