VWR® Signature™ Pipette Tips, 300 µl
Supplier: VWR International
Certificates
About this item
Clear, pretested polypropylene resin tips feature a uniquely thin and flexible top that conforms to the pipettor barrel, forming a tight seal with minimal ejection force.
- Sterile tips do not feature graduations
- Tips and hinged racks are autoclavable
- Available sterile or non sterile, with or without graduations
The unbeveled point is extremely precise, allowing you to easily dispense the smallest sample. A beveled mounting port ensures easy mounting for multichannel pipettors. Non sterile tips are graduated at 10, 20, 50, 100, 200 and 300 µl.
Certifications: Lot-certified pure and free of human DNA, DNase, RNase, endotoxins, and heavy metals.
Ordering information: Pak Rak® Express refills contain pipette tips to fill 8×12 racks, without the packaging waste of standard tip boxes. Pak Rak® Express refills require the Pak Rak® system loader (see 30128-374 series), which dispenses tips quickly and easily from the refill pack into each rack, eliminating risk of contamination from hands or gloves.