Nunc® 1536-Well Plates, Thermo Scientific
Catalog # 37001-002
Supplier: Thermo Fisher Scientific
Specifications
- Sterility:Non sterile
- Color:White
- Environmentally Preferable:
- Cat. No.:37001-002
- Packaging:30/Sleeve
- Supplier no.:253607
Specifications
About this item
These 1536-well plates are specifically designed for automation and ultra-high throughput screening
The MaxiSorp™ surface is recommended for assays where glycoproteins, such as antibodies, are adsorbed in the plastic. MaxiSorp™ surface has a a guaranteed affinity for proteins based on IgG adsorption.
They feature rounded square wells with flat bottoms to optimize dispenser and reader access. Plates have a total volume of 13µL per well and a working volume of 1–10µL per well. The center-to-center well distance is 2.25mm with a well depth of 5.0mm. The plates fit into a standard 96-well footprint compatible with standard equipment and automated handling systems. The skirted plate rim allows labeling and bar coding.
Ordering information: Sterile plates include a lid.