- Temperature range:0...100 °C
- Length:120 mm
- Material:Glass
- Environmentally Preferable:
- Cat. No.:97001-688
- Internal reference:ARGENTHAL™
- Junction:Ceramic
- Connector:S7
- Supplier no.:51343204
- Electrolyte:3M KCl
Electrode measures oxidation/reduction power of a solution
- Gold ring indicator
- Safelock™ closing ring
- ARGENTHAL™ lead-off and silver ion trap
- S7 screw head connector
Features a gold ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.