About this item
Revco® High-Performance Laboratory Freezers are designed for medical and scientific applications requiring storage of reagents, pharmaceuticals, biologicals, and other commonly used laboratory materials such as primers, siRNA molecules and FBS. A range of models offer cabinet sizes to fit specific laboratory space needs and storage volume requirements.
- Temperature is factory preset at -20 ° or -30 °C
- Capacities from 4.9 to 51.1 cu. ft. (139 to 1447 L)
- Icon-based controls and displays
- Large, easy-to-read digital display
- Graphic thermometer
Units are available in both automatic and manual defrost options
Heavy-duty refrigeration system is equipped with a hermetically sealed compressor. High-density, CFC-free urethane foam insulation minimizes the effects of ambient temperature fluctuations, reducing operating costs. A graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, and flashes bulb for undertemperature.
All freezers feature scratch-resistant, cold-rolled steel interior and exterior finishes. Doors on all units are self-closing and feature keyed locks. All models feature full length door handles, casters, and adjustable leveling feet.
Audible and visual warnings for overtemperature, undertemperature, and power failure. Alarm silence, ringback, and automatic reset functions are standard. Push-button alarm test and backup battery assure system readiness. Remote alarm contacts are standard on all models.