ContactUsLinkComponent
NeuroCult™ SM1 Medium without Vitamin A
NeuroCult™ SM1 Medium without Vitamin A
Catalog # MSPP-100-1282
CAS Number:  
undefined
NeuroCult™ SM1 Medium without Vitamin A
Catalog # MSPP-100-1282
Supplier Number:  100-1282
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    NeuroCult™ SM1 without vitamin A
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-100-1282
Specifications
About this item
Supplement (50X) without vitamin A for the culture of neural progenitor cells.
NeuroCult™ SM1 Without Vitamin A is a serum-free culture supplement, based on the published B27 formulation, with vitamin A removed. Vitamin A, also known as retinol, induces the differentiation of neural stem cells. In combination with a basal medium of choice, NeuroCult™ SM1 Without Vitamin A can be used for primary and pluripotent stem cell (PSC)-derived neural progenitor cell expansion and in customized differentiation protocols where vitamin A is not desired.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').