- Description:EpiCult™ Plus medium
- Size:500 ml
- Environmentally Preferable:
- Cat. No.:MSPP-06070
For culture of human, mouse, and rat epithelial stem cells.
EpiCult™ Plus Medium is a serum- and BPE-free culture medium for the expansion of epithelial stem cells from mammary, prostate, and other stratified and pseudostratified epithelial tissues from human, mouse, and rat. Epithelial cells can be expanded in EpiCult™ Plus Medium for multiple passages and for <30 population doublings; for some human mammary cells, addition of fetal bovine serum may be required. EpiCult™ Plus Medium is not recommended for human keratinocytes or primary human airway cells; for culturing primary human airway cells, we recommend PneumaCult™-Ex Plus Medium.
Note: Hydrocortisone stock solution is required for preparation of complete EpiCult™ Plus Medium.