ContactUsLinkComponent
96-Well Deep Well Plate, 0.5 ml, V Conical Bottom, Square Well, Sterile
96-Well Deep Well Plate, 0.5 ml, V Conical Bottom, Square Well, Sterile
Catalog # 104027-568
Supplier:  Alkali Scientific
undefined
96-Well Deep Well Plate, 0.5 ml, V Conical Bottom, Square Well, Sterile
Catalog # 104027-568
Supplier:  Alkali Scientific
Supplier Number:  TL456
Specifications
  • Sterility:
    Sterile
  • Volume:
    0.5 ml
  • Description:
    96-well deep well plate, square well
  • Environmentally Preferable:
  • Cat. No.:
    104027-568
  • Packaging:
    100/case
  • For use with:
    For KingFisher extraction platform
  • Bottom style:
    V connical bottom
Specifications
About this item
Conduct efficient sample processing with CellPro™ 0.5 ml 96-well deep well plate, featuring a V conical bottom and square well design, suitable for the KingFisher extraction platform, and available sterile in a pack of 100.
  • Made of medical grade polypropylene
  • Compatible with Thermo Fisher KingFisher™ instruments
  • Low binding assures maximum recovery
This 0.5 ml 96-well deep well plate, designed for use with the KingFisher extraction platform. Featuring a V conical bottom and square wells, these plates ensure efficient sample recovery and processing. Manufactured under sterile conditions, they are ready for immediate use in your laboratory workflows. With a capacity of 0.5 ml per well, these plates provide ample space for sample handling, making them ideal for high-throughput applications such as nucleic acid extraction, PCR setup, and sample storage.