ContactUsLinkComponent
96 Well White Plates with Clear Bottom, PS/PS, TC treated
96 Well White Plates with Clear Bottom, PS/PS, TC treated
Catalog # 104027-716
Supplier:  Alkali Scientific
undefined
96 Well White Plates with Clear Bottom, PS/PS, TC treated
Catalog # 104027-716
Supplier:  Alkali Scientific
Supplier Number:  TPW0096
Specifications
  • No. of wells:
    96
  • Sterility:
    Sterile
  • Color:
    White
  • Description:
    Cell culture plates, TC treated
  • Material:
    PS
  • Environmentally Preferable:
  • Cat. No.:
    104027-716
  • Packaging:
    Sterile 5/25
Specifications
About this item
Perform cell-based assays with accuracy using the Alkali Scientific™ 96 well white plate with clear bottom, TC treated for optimal cell attachment, supplied sterile in packs of 5 inner packs and cases of 25.
  • White wall and topside (PS)/clear bottom
  • Bottom material: Optically clear PS
  • Designed to prevent cross contamination
  • Effective gas exchange lid inner design
  • Alphanumeric labeling
Experience enhanced signal amplification and minimized cross-talk in your luminescence assays with 96-well white plates featuring a clear bottom. Crafted from premium polystyrene (PS/PS) and treated for tissue culture (TC), these plates are sterile and optimized for precise cell attachment and proliferation. The highly transparent bottom ensures optimal microscopic observation, while the white wall and topside minimize background interference. Each pack contains 5 plates, individually sterilized to prevent cross-contamination and labeled with alphanumeric markings for easy identification. With effective gas exchange lid inner design and assurance of non-pyrogenic, non-cytotoxic, DNase/RNase-free, and human DNA-free characteristics, these plates offer a reliable platform for your cell culture research needs. Trust in the quality and performance of our 96-well white plates to elevate your experimental outcomes with confidence.