ContactUsLinkComponent
ProCycle™ 0.2 ml Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
ProCycle™ 0.2 ml Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Catalog # 104027-482
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.2 ml Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Catalog # 104027-482
Supplier:  Alkali Scientific
Supplier Number:  PC7061
Specifications
  • Color:
    Clear
  • Description:
    0.2 ml rigid stable 8-Strip PCR tubes with attached individual flat caps
  • Environmentally Preferable:
  • Cat. No.:
    104027-482
  • Packaging:
    120×8, 960 per pack
Specifications
About this item
Ensure stability in PCR applications with Alkali Scientific™ ProCycle™ 0.2 ml rigid stable 8-Strip PCR tubes with attached individual flat caps, providing 120 strips per pack.
  • 0.2 ml 8-Strip PCR tubes with attached caps (120 strip×8, 960/pk)
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Optical flat caps
  • Certified RNAse, DNAse and pyrogen free
The StableStrip™ 0.2 ml 8-Strip PCR tube incorporates individually attached caps (flat or domed) for more convenience. No need to carry two separate components in inventory. The strip includes 8 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with either flat (needle pierceable) or dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the whole thermal cycle. The cap has an integral seal preventing contamination with surface of lid.
While more easily handled than single tubes, the 8-Strip PCR tubes will precisely fit standard well spacing and can also be used with multi-channel hand-held pipettors. Manufactured under the most stringent conditions to attain the highest quality standards in the industry.
Tubes are made of ultra-clear polypropylene plastic and strips can be cut to any length.