Nalgene® Round-Bottom Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Strong, crystal clear polycarbonate tubes for use with temperatures from –135 to 135 °C (–211 to 275 °F).
- For use in refrigerated and non-refrigerated rotors
- Non-toxic
- Autoclavable
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Special molding technique provides uniform wall thickness for greater strength.
Ordering information: Supplied without closures. Polypropylene closures available separately (see 21010-852 series).
Frequently Bought Together