PAXgene™ Blood RNA Tube, BD Biosciences
Catalog # 77776-026
Supplier: BD
Specifications
- Description:PAXgene™ Blood RNA Tube
- Environmentally Preferable:
- Cat. No.:77776-026
- Supplier no.:762165
Specifications
About this item
The PAXgene™ Blood RNA tube is part of a system that standardizes and integrates the key steps of whole blood collection, nucleic acid stabilization, and RNA purification
By minimizing the unpredictability of RNA processing, the system provides increased accuracy of intracellular RNA analysis. Tubes contain a proprietary reagent that immediately stabilizes intracellular RNA for three days at 18 to 25°C (64 to 77°F) and five days at 2 to 8°C (35 to 46°F). The ability to standardize the preanalytical process greatly enhances test accuracy and lab efficiency. Tube size is 16x100mm, with a capacity of 2.5 mL. Tubes feature a clear BD Hemogard™ closure and paper label for marking.
Certifications: CE marked.