Order Entry
United States
ContactUsLinkComponent
 
Thiamine (Vitamin B1) solution 1%, CellPro™
Catalog # 104028-966
Supplier:  Alkali Scientific
CAS Number:  
undefined
Thiamine (Vitamin B1) solution 1%, CellPro™
Catalog # 104028-966
Supplier:  Alkali Scientific
Supplier Number:  DS0197
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    104028-966

Specifications

About this item

Synonyms: Thiamine monochloride, Aneurine, Vitamin B1, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride
 :
 :300.81
UN:3248
CAS Number:59-43-8
Formula:C₁₂H₁₇ClN₄OS
MDL Number:MFCD00044586
Density:1.43
Merck Index:12,09430