Order Entry
United States
ContactUsLinkComponent
Pierce™ Mitochondria Isolation Kit for Tissue, Thermo Scientific
Pierce™ Mitochondria Isolation Kit for Tissue, Thermo Scientific
Catalog # PI89801
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Mitochondria Isolation Kit for Tissue, Thermo Scientific
Catalog # PI89801
Supplier:  Invitrogen
Supplier Number:  89801
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mitochondria Isolation Kit for Tissue
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    PI89801
  • Application:
    Intact Mitochondria from Mammalian Tissue
  • Supplier no.:
    89801
  • Sufficient for:
    50 Samples each containing 50-200 mg of Soft or Hard Tissue

Specifications

About this item

The Mitochondria Isolation Kit for Tissue enables isolation of intact mitochondria from soft and hard tissue samples.

  • Quick and convenient - isolate intact mitochondria in less than 60 minutes
  • Versatile - offers two methods of isolation from both soft and hard tissue samples
  • Multi-sample format - reagent-based approach enables simultaneous processing of multiple samples
  • Optional alternate method - reagents and protocol included for the traditional Dounce homogenization procedure with fewer required strokes
  • Bench-top compatible - both procedures can be performed using a microcentrifuge tube

Both reagent based and Dounce homogenization procedures use differential centrifugation to separate the intact mitochondria using a bench top microcentrifuge and are completed in less than 60 minutes. In addition, both procedures have been optimized for maximum yield of mitochondria with minimal damage to integrity. The isolated mitochondria may be used for many downstream applications, including 1D and 2D western blotting and protein profiling using mass spectroscopy.

Delivery: Kit contains Mitochondria isolation reagent A (50 mL), Mitochondria isolation reagent B (0.5 mL), Mitochondria isolation reagent C (65 ml), Bovine Serum Albumin (235 mg) and PBS Pack (makes 500 ml), 1 pack.