ContactUsLinkComponent
Analytical Mill, A 10 basic
Analytical Mill, A 10 basic
Catalog # 76329-858
Supplier:  IKA Works
undefined
Analytical Mill, A 10 basic
Catalog # 76329-858
Supplier:  IKA Works
Supplier Number:  0025002269
Specifications
  • Description:
    A 10 basic
  • Environmentally Preferable:
  • Cat. No.:
    76329-858
Specifications
About this item
This batch mill has simple handling with keypads which grinds hard, brittle, soft and fibrous materials, because samples may be embrittled directly in the grinding chamber- tough, oily and aqueous samples can also be grinded. Easily exchangeable beater/cutter for a variety of applications.
  • Counter- displays grinding time and error code
  • Digital timer with interval function, integrated cooling connections
  • Brushless motor for long service life and low noise level
  • Bayonet lock for lid to enable simple and safe locking
The mill starts only when the lid is closed and can be opened at standstill. A quick stop feature further increases the safety of the user.