ContactUsLinkComponent
SMD-VAC™-HP Vacuum Handling System, Virtual Industries
Catalog # 21925-200
Supplier:  Virtual Industries
undefined
SMD-VAC™-HP Vacuum Handling System, Virtual Industries
Catalog # 21925-200
Supplier:  Virtual Industries
Supplier Number:  VFS-1
Specifications
  • Description:
    Optional Foot Switch
  • Environmentally Preferable:
  • Cat. No.:
    21925-200
  • Supplier no.:
    VFS-1
Specifications
About this item
Made with extra power to pick up very small to very large-size components, the SMD-VAC™-HP features a precision pump and motor that generates over 15"Hg
Compact tower design has a small footprint. Electric eye pen holder automatically shuts the unit down when pen is returned to holder. Optional foot switch 21925-200 controls vacuum during pick-and-place operations.
Ordering information: Complete with 1.2m (4') of coiled and straight vacuum hose, a vacuum pick-up handle, set of seven vacuum tips, 12V DC wall transformer, automatic pump on/off control, rubber nonskid feet and ESD connection.