ContactUsLinkComponent
Surgical Instrument Cleaner and Lubricant, Integra™ Miltex®
Surgical Instrument Cleaner and Lubricant, Integra™ Miltex®
Catalog # 95042-720
Supplier:  Miltex
undefined
Surgical Instrument Cleaner and Lubricant, Integra™ Miltex®
Catalog # 95042-720
Supplier:  Miltex
Supplier Number:  3-710
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    3.8 L (1 gal.)
  • Environmentally Preferable:
  • Cat. No.:
    95042-720
  • Supplier no.:
    3-710
Specifications
About this item
Dual-purpose instrument cleaner and lubricant is chemically engineered for use as a presoak or for manual cleaning in washers, decontaminators, ultrasonic cleaners, and cart washers
The biodegradable solution ensures cleaner and brighter instruments, and will remove stains, blood, and mineral deposits.