Human Recombinant DPP7/DPPII/DPP2 Protein (His Tag) (From HEK293 Cells)
Catalog # 77446-782
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Environmentally Preferable:
- Tag sequence:His Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 ℃. Store it under sterile conditions at −20 to −80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:DPP2,DPPII,QPP
- Protein/Peptide Name:DPP7
- Purity:≥90% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant human DPPII comprises 482 amino acids with a predicted molecular mass of 53.6 kDa. As a result of glycosylation, rh DPPII migrates as an approximately 60 kDa band in SDS-PAGE under reducing conditions.
- Shipping Temperature:Ambient
- Cat. No.:77446-782
Specifications
About this item
A DNA sequence encoding the human DPPII (NP_037511.2) (Met 1-Leu 492) with a C-terminal polyhistidine tag was expressed.