Liebig Condenser, Chemglass
Catalog # 80067-368
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Liebig Condenser
- Environmentally Preferable:
- Cat. No.:80067-368
- Supplier no.:CG-1218-01
Specifications
About this item
Condenser has a [ST] outer joint at the top and a lower inner drip tip joint
Hose connections have an O.D. of 10mm at the middle serration. [ST] joint: 24/40.