FLAMEBOY™ Sterilizing Pistol, INTEGRA Biosciences
Supplier: Avantor
Certificates
About this item
The FLAMEBOY™ is a handy and portable flame sterilizer with piezo-electric ignition for safe flame sterilization
Alternatively, the FLAMEBOY™ can be connected to a gas distribution system for use in a permanent installation, such as hoods and laboratory environments.
Equipped with a gas cartridge adapter for Camingaz CV360, the FLAMEBOY™ operates independently of any gas distribution system. As a result, it is conveniently at hand wherever needed, be it production plant, fermenter, laboratory, or outdoors. The gas cartridge adapters are easily mounted via an intelligent "click and go" mechanism.
Ordering information: Includes 3 nozzles for propane/butane, town gas, and natural gas. Also includes adapter for standard gas hose and threaded adapter for ¼" gas hose.
Frequently Bought Together