SARS-CoV Recombinant Spike RBD Protein (RBD, mFc Tag) (from HEK293 Cells), Biotinylated
Catalog # 77447-744
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS
- Size:20 µg
- Environmentally Preferable:
- Tag sequence:RBD, mFc Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20...−80 °C. Store it under sterile conditions at −20...−80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:novel coronavirus RBD|novel coronavirus s2|ncov RBD|ncov s2|ncov spike|coronavirus s2|S1|novel coronavirus s1|novel coronavirus spike|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|s2|RBD
- Protein/Peptide Name:Spike
- Purity:>90 % as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV Spike/RBD Protein (RBD, mFc Tag) consists of 457 amino acids and predicts a molecular mass of 51.49 kDa. As a result of glycosylation, it migrates as an approximately 58.01 and 60.51 KDa band in SDS-PAGE under reducing conditions.
- Shipping Temperature:Ambient
- Cat. No.:77447-744
Specifications
About this item
A DNA sequence encoding the receptor binding domain (RBD) of SARS-CoV (isolate:WH20) spike (AAX16192.1) (Arg306-Phe527) was expressed with the Fc region of mouse IgG1 at the C-terminus. The purified protein was biotinylated in vitro.