Order Entry
United States
ContactUsLinkComponent
Rat TGFb1 (Transforming Growth Factor Beta 1) ELISA Kit
Rat TGFb1 (Transforming Growth Factor Beta 1) ELISA Kit
Catalog # 77514-048
Supplier:  AFG Bioscience
New Product
CAS Number:  
undefined
Rat TGFb1 (Transforming Growth Factor Beta 1) ELISA Kit
Catalog # 77514-048
Supplier:  AFG Bioscience
Supplier Number:  EK241816
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Rat
  • Primary antibody reactivity:
  • Target protein:
    TGFb1 (Transforming Growth Factor Beta 1)
  • Size:
    96 wells
  • Environmentally Preferable:
  • Sample Type:
    Serum, platelet-poor plasma, tissue homogenates, cell culture supernatants and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    15.63 - 1000 pg/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Cytokine, tumor immunity, infection immunity
  • Sensitivity:
    6.4 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77514-048

Specifications

About this item