About this item
Clear lid with large horizontal opening accommodates larger syringes, allowing one-hand, one-step, no-touch sharps depositing
GatorGuard™ containers feature a tumbler design, capacity indicator, lift tabs to assist deposit, and horizontal drop lid to maximize container volume. The tumbler design allows disposal of various sized items, from a lightweight butterfly catheter to a 60cc syringe. Tortuous path containers capture disposed items in a winding channel inside the container.
All Sharps-A-Gator® containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Anti-spill inner flap on the lid impedes hand entry. Container base has hand grips for easy transport. Containers lock for final disposal. Plastics and pigments are safe to autoclave or incinerate. Containers can be placed on countertops or mounted on a wall.