Screen Prep™ Presaturated Wipes, ITW Chemtronics®
Catalog # 47733-170
Supplier: ITW Chemtronics
Specifications
- Description:Screen Prep™ Wipes, Twinpacks
- Environmentally Preferable:
- Cat. No.:47733-170
- Supplier no.:CSP20
Specifications
About this item
A dry and wet wipe cleaning and static control system.
- Dry wipes optically clean uncoated monitor screens
- Safely remove dust, dirt, and oils
Wet wipes provide a static dissipative treatment with the convenience of a disposable applicator. Control static-generated contamination. Ideal for field service kits. Each dry wipe and wet wipe is hermetically sealed to prevent evaporation and maintain solvent effectiveness, then folded together in a twinpack. (ITW Chemtronics CSP20)