Specifications
- Color:Silver
- Description:Stainless steel binClip for stainless steel LocBoard®, 3 per pack
- Environmentally Preferable:
- Cat. No.:77269-322
Specifications
About this item
The stainless steel LocHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
- Position hanging plastic bins individually anywhere on a stainless steel LocBoard
- plastic hanging bins mount on top of the BinClip which secures the bin to the LocBoard
- Ideal for use everywhere from clean rooms to harsh environments
- Use multiple stainless steel BinClips for larger bins and heavier items
- Won't rust or corrode in any environment, even ones with extreme temperatures
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and The stainless steel pegboard storage from Triton products provides the ideal answer. Two innovative best-selling products, the LocBoard and the LocHook, are now available in stainless steel. They provide a commercial grade heavy-duty pegboard and hooks system. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect stainless steel pegboard solution. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of the stainless steel pegboard system from Triton products.