2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
Boiling Pt:82
Melting Pt:-89
Density:0.786 g/cm³ (20 °C)
Molecular Weight:60.1 g/mol
Flash Pt:12
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3 II
Merck Index:13,05228
:Management of Change (MOC) category = R
REACH:01-2119457558-25
:GHS02
:GHS07
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (CH₃CHOHCH₃):≥ 99.5 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 4 ppm
- Solubility in H₂O:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤ 0.05 %
- Acidity (µeq/g):≤ 0.2
- Alkalinity (µeq/g):≤ 0.1
- Chloride (Cl):≤ 0.2 ppm
- Phosphate (PO₄):≤ 0.3 ppm
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Arsenic and Antimony (as As):≤ 10.0 ppb
- Trace Impurities - Barium (Ba):≤ 20.0 ppb
- Trace Impurities - Beryllium (Be):≤ 100.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 100.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 20.0 ppb
- Trace Impurities - Calcium (Ca):≤ 50.0 ppb
- Trace Impurities - Chromium (Cr):≤ 20.0 ppb
- Trace Impurities - Cobalt (Co):≤ 20 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Gallium (Ga):≤ 50 ppb
- Trace Impurities - Germanium (Ge):≤ 50.0 ppb
- Trace Impurities - Gold (Au):≤ 20 ppb
- Heavy Metals (as Pb):≤ 200.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 20.0 ppb
- Trace Impurities - Lithium (Li):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 20 ppb
- Trace Impurities - Manganese (Mn):≤ 15.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 100.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Niobium (Nb):≤ 100.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 50 ppb
- Trace Impurities - Silver (Ag):≤ 20.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 20.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 100.0 ppb
- Trace Impurities - Thallium (Tl):≤ 10.0 ppb
- Trace Impurities - Tin (Sn):≤ 100 ppb
- Trace Impurities - Titanium (Ti):≤ 20 ppb
- Trace Impurities - Vanadium (V):≤ 100.0 ppb
- Trace Impurities - Zinc (Zn):≤ 50 ppb
- Trace Impurities - Zirconium (Zr):≤ 100.0 ppb
- Particle Count - 0.2 µm and greater:≤ 5000 par/ml
- Particle Count - 0.3 µm and greater:≤ 5000 par/ml
- Particle Count - 0.5 µm and greater:≤ 50 par/ml
- Particle Count - 1.0 µm and greater:≤ 8 par/ml
Specification Test Results
Related Information
Certificates
Search by Lot Number9079 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT9079-5
Product specification
Frequently Bought Together