Order Entry
United States
ContactUsLinkComponent
 
Sodium thiosulfate
Catalog # RC7958-5
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Sodium thiosulfate
Catalog # RC7958-5
Supplier:  Ricca Chemical
Supplier Number:  R7958000-20F
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Cubitainer®
  • Size:
    20 L
  • Environmentally Preferable:
  • Cat. No.:
    RC7958-5
  • Supplier no.:
    R7958000-20F

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.1574 ± 0.0001 N at 20.0 °C

Specification Test Results

About this item

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite
 :
 :158.11 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
Boiling Pt:100
Melting Pt:48
Density:1.0200 g/cm³
Merck Index:12,08844