TSX Universal series ULT freezers are the pinnacle of reliability, sustainability, and innovation incorporating insights from over 80 years of experience to enhance every aspect of the system. Updates to the variable speed compressors has made the TSX Universal adaptable across all workflows. Available in four capacities, they are ENERGY STAR® certified with a 33% energy efficiency improvement, ACT labeled and produced in a Zero-Waste to Landfill certified facility.
- Universal V-Drive technology delivers tighter peak variation, faster recovery times, decrease in energy consumption, improved pull down time
- Improved user interface: Full 7" capacitive touchscreen
- Reduced rapid re-entry times: Improved pressure relief port enables re-entry in just 30 seconds helping ensure uninterrupted access to freezer
- Expanded setpoint range: −40 to −86 °C
- Universal voltage: Operates on 100 to 230 V, 50/60 Hz, streamlining procurement and qualification strategies from early research and development to global clinical trials
- Simplified updates: Onboard software streamlines startup and integration, making it easier to access and install the latest updates for continuous access to new features
- Industry-leading warranty: 12 years on compressor parts and five years unit
Whisper-quiet noise under 44 dBA allows the TSX Universal ULT to be placed anywhere without causing sound pollution. Connectivity integration solutions helps monitor, share, and analyse data allowing streamlined processes and improved research outcomes.
By incorporating environmental sustainability principles into each design step, the environmental impact of these products is reduced early in the design process, helping to deliver on our commitment to achieve net-zero emissions by 2050 while helping customers advance their sustainability goals. Our strategy focuses on five key impact areas: Less hazardous, less waste, more energy efficient, responsibly packaged and extended life. All TSX Universal ULTs beat today’s ENERGY STAR criteria by wide margins and positioned to meet tightening requirements in the future.
Choice of four capacities: 400, 500, 600 and 700 (50.8 mm box)
- Catalog No:
- 77400-778
- 77400-776
- 77400-774
- 77400-772
- 77400-790
- 77400-788
- 77400-806
- 77400-786
- 77400-804
- 77400-784
- 77400-802
- 77400-782
- 77400-780
- 77400-824
- 77400-822
- 77400-820
- 77400-800
- 77400-818
- 77400-798
- 77400-816
- 77400-796
- 77400-814
- 77400-834
- 77400-794
- 77400-812
- 77400-832
- 77400-792
- 77400-810
- 77400-830
- 77400-808
- 77400-828
- 77400-826
- Model:
- TSX40086FARAKCO
- TSX40086FARAK
- TSX40086FALB
- TSX40086FA
- TSX50086FALB
- TSX40086FA
- TSX60086FALB
- TSX40086LA
- TSX40086FA
- TSX40086FARAKLN
- TSX50086LA
- TSX40086FARAKLB
- TSX40086FARAKHID
- TSX70086FARAK
- TSX70086FALB
- TSX40086FA
- TSX50086FARAKLN
- TSX60086LA
- TSX50086FARAKLB
- TSX60086FARAKLN
- TSX50086FARAKHID
- TSX60086FARAKLB
- TSX70086LA
- TSX50086FARAKCO
- TSX60086FARAKHID
- TSX70086FARAKLN
- TSX50086FARAK
- TSX60086FARAKCO
- TSX70086FARAKLB
- TSX60086FARAK
- TSX70086FARAKHID
- TSX70086FARAKCO
- Defrost:
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Electrical:
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
- 100 - 230 V, 50/60 Hz
TDS Thermo Scientific Ultra-Low Temperature Freezer, TSX40086FA TDS Thermo Scientific Ultra-Low Temperature Freezer, TSX40086LA TDS Thermo Scientific Ultra-Low Temperature Freezer, TSX50086FA TDS Thermo Scientific Ultra-Low Temperature Freezer, TSX50086LA TDS Thermo Scientific Ultra-Low Temperature Freezer, TSX60086FA