Brix Hydrometers with Thermometer (°C)
Supplier: Heathrow Scientific
Certificates
About this item
Our brix scale hydrometers with °C thermometer are calibrated to show the percentage of sucrose by weight at 20 °C.
- Calibrated to show percent sucrose by weight
- Wide variety of brix ranges available
- Thermometers contain safety blue liquid fill
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
The thermometer scale is located in the body and features safety blue liquid fill. The thermometers have a range of 0 to 50 °C with 1° subdivisions. A correction scale for conversion of reading to 20 °C is printed opposite of the temperature scale.
Frequently Bought Together