Digital Refractometer, Seawater/Salinity, 4 Scales, MDX-202
76545-888
- Resolution:Salinity: 1 ppt
Chlorinity: 1 ppt
Specific gravity: 0.001
Refractive index: 0.0001 nD - Environmentally Preferable:
- Cat. No.:76545-888
- Accuracy:Salinity: ±2 ppt
Specific gravity: ±0.002
Refractive index: ±0.0003 nD - Scale range:Salinity: 0 - 100 ppt
Chlorinity: 0 - 57 ppt
Specific gravity: 1.000 - 1.075
Refractive index: 1.3330 - 1.4200 nD
The MDX-Series delivers digital accuracy and high-end features for specific applications. Digital seawater refractometers can be used to measure salinity in the aquatics industry.
- Compact and lightweight
- Removable grip and prism cover for easy cleaning
- Continuous loop read features (15 reads)
- Automatic temperature control
- Zero with distilled water
- 589.3 nm LED
- Stainless steel prism well
- °C/°F switchable temperature display
- 1 year warranty
The MDX-Series Digital Brix/Sugar Refractometer features IP 65/67 ratings making it ideal for use in the field and in a multitude of environments. Delivering quick, digital accuracy with high-end performance features such as Automatic Temperature Compensation (ATC) and Continuous Loop Average Readings.
: IP protection class according to DIN EN 60529: IP 67 (for detecting unit) and IP 65 for remainder.
: Supplied with digital refractometer (Model MDX-202), rubber grip and prism cover, hard shell case with protective lining, disposable pipet, battery and user manual.