Diethyl ether stabilised USP for anesthesia, Macron Fine Chemicals™
Catalog # MK080410
Supplier: Avantor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Aluminum bottle
- Size:500 g
- Environmentally Preferable:
- Cat. No.:MK080410
- Supplier no.:0804-10
Specifications
Specification Test Results
- USP - Specific Gravity at 25°/25°C:0.713 - 0.716
- USP - Acidity (as CH₃COOH):≤0.003 %
- USP - Aldehydes:Passes Test
- USP - Low-Boiling Hydrocarbons:≤0.2 %
- USP - Nonvolatile Residue:≤0.003 %
- USP - Peroxide (as H₂O₂):≤0.3 ppm
- USP - Preservative (BHT):≥0.00005 %
- USP - Water (H₂O)(by Karl Fischer titrn):≤0.2 %
Specification Test Results
About this item
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether
:
:74.12 g/mol
Flash Pt:–40 °C
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
Boiling Pt:35
Melting Pt:-123
Density:0.7135 g/cm³ (20 °C)
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
ADR:3 I
Merck Index:12,03852
:Management of Change (MOC) category = R