Dichloromethane ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Supplier: Honeywell Research Chemicals
Certificates
About this item
Synonyms: Methane dichloride, Methylene chloride, Methylene chloride
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
Boiling Pt:39.8
Density:1.33 g/cm³
:Stabiliser
Molecular Weight:84.93 g/mol
Melting Pt:-95
Flash Pt:Does not flash
UN:1593
ADR:6.1 III
Merck Index:12,06140
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Passes test
- Color (APHA):10 max.
- Free Halogens:Passes test
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:3 mg/L max.
- Titrable Acid:0.3 µeq/g max.
- UV Cutoff:233 nm max.
- Water by Karl Fisher Titration:0.02 % max.
Specification Test Results
Related Information
Certificates
Search by Lot NumberHB Honeywell B
Datasheet
VWR GHS Compliance Letter (US)
Additional Product Details
VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Frequently Bought Together