VWR® Clip-It Timer™ Timer
Catalog # 62379-606
Supplier: VWR International
Specifications
- Environmentally Preferable:
- Cat. No.:62379-606
- W×H:3.8×5.1 cm (1.5×2)"
Specifications
About this item
Pocket-size timer performs countdown, alarm, and stopwatch functions and can be used to catch lab test results, signal drying/incubating times, or as alarm for appointments.
- Clips onto lab coat, notebook, or clipboard
- Traceable®
- Housed in a rugged, chemical-resistant ABS case
Features quartz-crystal accuracy and easy-to-use three-button operation. Four-digit 8 mm (⁵/₁₆") high LCD displays time remaining in hours and minutes. Countdown mode programmable from 100 hours to 1 second. Stopwatch mode times from 1 second to 99 hours and then repeats itself. Timer also has time-out feature in both up and down counting. Loud beeping alarm sounds for 60 seconds.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with magnet for placing on metal and an 18-month continuous-use, 1.5 V silver-oxide battery.