Order Entry
United States
ContactUsLinkComponent
Weller® UNGAR® Flux Solvent Dispenser, Cooper Tools
Weller® UNGAR® Flux Solvent Dispenser, Cooper Tools
Catalog # 37695-400
Supplier:  Cooper Tools
undefined
Weller® UNGAR® Flux Solvent Dispenser, Cooper Tools
Catalog # 37695-400
Supplier:  Cooper Tools
Supplier Number:  FD2

Specifications

  • Capacity:
    60 ml (2 oz.)
  • Environmentally Preferable:
  • Cat. No.:
    37695-400
  • Needle length:
    0.5 mm (0.020")
  • Supplier no.:
    FD2

Specifications

About this item

Avoids spills and waste and places flux precisely where needed.

Simply squeeze bottle to dispense one drop of liquid resin, flux, or oil solvent through stainless steel needle.