Agfa® X-Ray Film
Supplier: Associated Metals
Certificates
About this item
High quality general radiographic film may be used with any medical X-ray machine for routine X-rays in primary medical procedures
Definition is medium contrast. Blue sensitive film is available in high speed 800 or half speed 400. Speeds are dependent upon screen being used. Store in cool, dry place.
Ordering information: Film is packaged 100 sheets per pack.
Frequently Bought Together