Specifications
- Shoe size:One size
- Garment application:
- Gender:
- Risk area:
- Color:Transparent
- Environmentally Preferable:
- Cat. No.:77395-160
Specifications
About this item
A kit consisting of sterile woven cotton slippers inserted in a sterile bag. The kit allows for large area sampling at ground level.
- Ideal for sample collection and inspection of surfaces in contact with livestock
- Easy to use and highly versatile
- Simultaneous sample collection and inspection
- Sterile and biocide-free
- Fits any work boots size
Wipa-boots are impregnated with a solution that allows transportation of different pathogens. A major concern in the livestock industry is the prevention, detection, and control of infections by pathogens like viruses (H1N1, African swine fever, PRRS, Circovirus, Newcastle) and bacteria (Salmonella and E. coli including STEC). This product is a quick and simple solution to establish an innocuous environment in livestock housing, animal transportation vehicles, and their washing stations.
Acting as a quick and simple sterile sampling method, Wipa-boots allow sample collection to occur simultaneously during routine walking inspections. The kit includes a sterile bag and a pair of boot covers, pre-moistened in 23 ml of tryptone salt. This solution allows the transportation of different pathogens, for reliable and safe analysis.