ContactUsLinkComponent
Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Catalog # 55709-903
undefined
Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Catalog # 55709-903
Supplier Number:  CS509X3
Specifications
  • No. of Holes:
    25
  • Description:
    For Locator 4
  • Dimensions:
    76×76 mm (3×3")
  • Environmentally Preferable:
  • Cat. No.:
    55709-903
  • Supplier no.:
    CS509X3
Specifications
About this item
Nalgene® Cryoplastic boxes are of durable, ultra-low temperature polycarbonate construction
One-way lid reduces the chances of sample spillage. A printed grid system on the lid facilitates easy logging and retrieval of samples.