Ward's® Rotating Mixers and Incubators, 230 V
Supplier: VWR International
Certificates
About this item
Suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry and hybridization.
- 230 V model
- Includes tube holders for common tube sizes
- Several motion and incubation capacities available
- Two-year warranty
All mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5 to 50 ml. Motion on all units can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing.
The 'PLUS' units have the following additional functions: 'Mix' function: Rotating in combination with oscillations (increased mixing); 'Pause' function: Rotating with intermittent 'stops' during rotation; and 'Rock' function: Back and forth, gentle rocking motion (see-saw).
Caution: 230 V model. Corresponding model with voltage suitable for use in US (470313-910, 470313-912, 470313-914 and 470313-920) are also available, please see separate product page.
Frequently Bought Together