ContactUsLinkComponent
VWR® Dissecting Scissors, Curved Tip, 4¹/₂"
VWR® Dissecting Scissors, Curved Tip, 4¹/₂"
Catalog # 82027-584
Supplier:  VWR International
undefined
VWR® Dissecting Scissors, Curved Tip, 4¹/₂"
Catalog # 82027-584
Supplier:  VWR International
Specifications
  • Length:
    114 mm (4½")
  • Environmentally Preferable:
  • Cat. No.:
    82027-584
Specifications
About this item
VWR® Dissecting Scissors are stainless steel with fine, bent tips.
  • Stainless steel fabrication
  • Fine, curved tips
Dissecting scissors are used for a variety of applications and are available in different lengths.