Order Entry
United States
ContactUsLinkComponent
Stilla® EGFR 6-color Crystal Digital PCR® Kit
Stilla® EGFR 6-color Crystal Digital PCR® Kit
Catalog # 77392-296
CAS Number:  
undefined
Stilla® EGFR 6-color Crystal Digital PCR® Kit
Catalog # 77392-296
Supplier Number:  R30006
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    EGFR 6-color crystal digital PCR® kit
  • Environmentally Preferable:
  • Cat. No.:
    77392-296
  • Units:
    192 ruby reactions
  • Details:
    Storage temperature: −20... ±5 °C

Specifications

About this item

The EGFR 6-color crystal digital PCR® kit allows for the detection and quantification of up to 32 somatic EGFR mutations in exons 18, 19, 20, and 21 associated with non-small cell lung cancer (NSCLC).

  • Ready-to-use detection kit for EGFR mutations in exons 18, 19, 20, and 21
  • 48 reactions on the sapphire chip on the 6-color naica® system
  • 192 reactions on the ruby chip on the Nio™+
  • Sample matrix: cfDNA from plasma and DNA from FFPE tissue biopsies

The EGFR 6-color crystal digital PCR® kit provides a fully validated, specific, and ultrasensitive assay for the 6-color naica® system with the sapphire chip and Nio™+ with the ruby chip. The kit can be used to test cell-free DNA (cfDNA) extracted from plasma samples as well as DNA extracted from FFPE tissue biopsies.

The EGFR 6-color crystal digital PCR® kit is a ready-to-use solution comprised of six components, including the high-performance naica® multiplex PCR MIX 10X, an optimal primer and probe solution, a set of positive and negative controls, and water for sample dilution.

Delivery: Dry-ice shipment.
Caution: Store at −20 ±5 °C. For research use only. Not for use in diagnostic procedures.