- Environmentally Preferable:
- Cat. No.:76443-168
- Electrical:200 - 230 V
- Weight:4.5 kg
These digital oil baths are easy to use, they offer digital settingin a compact design oil bath.
- 200 - 230 V model, not for use in USA
- Fixed temperature operation function
- White LED digital display
- One year warranty
Exclusive connection for bath operational setting function with RE601/801 rotary evaporator, removable oil tank for convenient cleaning and changing of oil (75846-372). Flat-shaped bath with no heater or sensor inside makes for ease in cleaning (76443-166, 76443-168).
Safety device includes bath protection sheet, automatic overheat prevention, independent overheat prevention (fixed temperature), temperature upper limit error, overcurrent protection fuse; overshoot alert, auto resume (selectable), 2A service outlet, calibration offset (76443-166, 76443-168). Bath protection cover (ABS heat-resistant resin), self-diagnostic function (automatic overheat prevention, temperature sensor error, triac short circuit, heater disconnection, main relay failure), circuit protector, thermal fuse, micro switch to detect heating without oil (75846-372).